Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylbutyrolactone, Spectrum™ Chemical
SDP

Catalog No. 18620096 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-096 200 L
18-620-094 100 mL
18-620-095 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-096 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A2107200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 517-23-7
Molecular Weight (g/mol) 128.13

Specifications

Quantity 200 L
Packaging Drum
Molecular Formula C6H8O3
Boiling Point 107°C to 108°C
InChI Key OMQHDIHZSDEIFH-UHFFFAOYNA-N
SMILES CC(=O)C1CCOC1=O
IUPAC Name 3-acetyloxolan-2-one
Percent Purity 97%
Grade Ungraded
Chemical Name or Material 2-Acetylbutyrolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.