accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHOOHOOHOHOOOHO

CAS RN 517-60-2

IUPAC Name: benzene-1,2,3,4,5,6-hexacarboxylic acid
Synonyms: benzene hexacarboxylic acid benzenehexacarboxylate benzenehexacarboxylic acid hexasodium salt mellitic acid mellic acid
Molecular Weight (g/mol): 342.17
Molecular Formula: C12H6O12
InChi Key: YDSWCNNOKPMOTP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C(O)=O)C(C(O)=O)=C(C(O)=O)C(C(O)=O)=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results