Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mellitic Acid 98.0+%, TCI America™
SDP

Catalog No. B02465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02465G 5 g
B024625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02465G Supplier TCI America Supplier No. B02465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 517-60-2
Molecular Formula C12H6O12
Molecular Weight (g/mol) 342.168
MDL Number MFCD00002469
InChI Key YDSWCNNOKPMOTP-UHFFFAOYSA-N
Synonym Benzenehexacarboxylic Acid
PubChem CID 2334
ChEBI CHEBI:41089
IUPAC Name benzene-1,2,3,4,5,6-hexacarboxylic acid
SMILES C1(=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 288°C
Color White-Yellow
Quantity 5 g
Formula Weight 342.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mellitic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.