CAS RN 51716-96-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Brilliant Cresyl Blue Zinc Chloride Double Salt
Stains reticulocytes in blood and used for preparation of a staining solution