accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51751-16-7

IUPAC Name: 2-(2H-1,3-dithiol-2-ylidene)-2H-1,3-dithiole
Synonyms: tetrathiafulvalene delta(2),(2')-bi-1,3-dithiole TTF delta-2,2'-bi-1,3-dithiole delta-2:2'-bis(1,3-dithiazole) 1,4,5,8-tetrathiafulvalene
Molecular Weight (g/mol): 204.34
Molecular Formula: C6H4S4
InChi Key: FHCPAXDKURNIOZ-UHFFFAOYSA-N
SMILES: S1C=CSC1=C1SC=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results