Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrathiafulvalene-d4, TCI America™
SDP

Catalog No. T1377100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1377100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1377100MG Supplier TCI America Supplier No. T1377100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51751-16-7
Molecular Formula C6H4S4
Molecular Weight (g/mol) 208.362
MDL Number MFCD00191580
InChI Key FHCPAXDKURNIOZ-RHQRLBAQSA-N
Synonym TTF-d4
PubChem CID 12207880
IUPAC Name 4,5-dideuterio-2-(4,5-dideuterio-1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES C1=CSC(=C2SC=CS2)S1

Specifications

Melting Point 119°C
Quantity 100 mg
Formula Weight 208.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrathiafulvalene-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.