accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51787-96-3

IUPAC Name: 5-(4-fluorophenyl)oxolan-2-one
Synonyms: 5-(4-fluorophenyl)oxolan-2-one
Molecular Weight (g/mol): 180.18
Molecular Formula: C10H9FO2
InChi Key: RMFNZGXVLAUJHF-UHFFFAOYNA-N
SMILES: FC1=CC=C(C=C1)C1CCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results