Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-(4-Fluorophenyl)-γ-butyrolactone 98%, Thermo Scientific™

Catalog No. AC175230050
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC175230050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175230050 Supplier Thermo Scientific Chemicals Supplier No. 175230050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51787-96-3
Density 1.2
Infrared Spectrum Authentic
Molecular Formula C10H9FO2
Refractive Index 1.5216 to 1.5236 (20°C, 589nm)
Quantity 5g
MDL Number MFCD00005399
Synonym 4,5-dihydro-5-4-fluorophenyl-2 3h-furanone, 5-4-fluorophenyl oxolan-2-one, gamma-4-fluorophenyl-gamma-butyrolactone, 5-4-fluorophenyl dihydrofuran-2 3h-one, 2 3h-furanone, 5-4-fluorophenyl dihydro, .gamma.-4-fluorophenyl-butyrolactone, 5-4-fluorophenyl tetrahydrofuran-2-one, .gamma.-4-fluorophenyl-.gamma.-butyrolactone, 5-4-fluorophenyl-3,4,5-trihydrofuran-2-one, gamma-p-fluorophenyl-gamma-butyrolactone
InChI Key RMFNZGXVLAUJHF-UHFFFAOYSA-N
SMILES C1CC(=O)OC1C2=CC=C(C=C2)F
IUPAC Name 5-(4-fluorophenyl)oxolan-2-one
Molecular Weight (g/mol) 180.178
PubChem CID 103541
Formula Weight 180.18
Percent Purity ≥97.5%
Chemical Name or Material γ-(4-Fluorophenyl)-γ-butyrolactone, 98%
Show More Show Less
EINECSNumber 257-422-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.