accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOOOO

CAS RN 518-51-4

IUPAC Name: disodium 4-[1-(4-oxidophenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]benzen-1-olate
Synonyms: phenolphthalein sodium
Molecular Weight (g/mol): 362.29
Molecular Formula: C20H12Na2O4
InChi Key: GCZXTTJWUAJLNQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)