Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Disodium Salt 95%, Thermo Scientific™

Catalog No. AC417210250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC417210250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC417210250 Supplier Thermo Scientific Chemicals Supplier No. 417210250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 518-51-4
Assay Percent Range 95%
Molecular Formula C20H12Na2O4
MDL Number MFCD00051414
Merck Index 12, 7393
Infrared Spectrum Authentic
Formula Weight 362.28
Loss on Drying 9% max. (1 g, 105°C, 3 hrs)
Grade Extra Pure
UV_Vis 94% min. (on dry substance)
Color Brown to Purple
Quantity 25g
Chemical Name or Material Phenolphthalein disodium salt, 95%
Show More Show Less
EINECSNumber 208-254-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.