CAS RN 518-67-2
Dimidium bromide, 95%
CAS: 518-67-2 Molecular Formula: C20H18BrN3 Molecular Weight (g/mol): 380.29 MDL Number: MFCD00011757 InChI Key: MQOKYEROIFEEBH-UHFFFAOYSA-N Synonym: dimidium bromide,trypadine,3,8-diamino-5-methyl-6-phenylphenanthridinium bromide,phenanthridinium 1553,phenanthridinium compound 1553,phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide,2,7-diamino-10-methyl-9-phenylphenanthridinium bromide,3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide,phenanthridium 1553 PubChem CID: 68207 IUPAC Name: 5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide SMILES: [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Dimidium bromide, 98%
CAS: 518-67-2 Molecular Formula: C20H18BrN3 Molecular Weight (g/mol): 380.29 MDL Number: MFCD00011757 InChI Key: MQOKYEROIFEEBH-UHFFFAOYSA-N Synonym: dimidium bromide,trypadine,3,8-diamino-5-methyl-6-phenylphenanthridinium bromide,phenanthridinium 1553,phenanthridinium compound 1553,phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide,2,7-diamino-10-methyl-9-phenylphenanthridinium bromide,3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide,phenanthridium 1553 PubChem CID: 68207 IUPAC Name: 5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide SMILES: [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Dimidium Bromide 98.0+%, TCI America™
CAS: 518-67-2 Molecular Formula: C20H18BrN3 Molecular Weight (g/mol): 380.29 MDL Number: MFCD00011757 InChI Key: MQOKYEROIFEEBH-UHFFFAOYSA-N Synonym: dimidium bromide,trypadine,3,8-diamino-5-methyl-6-phenylphenanthridinium bromide,phenanthridinium 1553,phenanthridinium compound 1553,phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide,2,7-diamino-10-methyl-9-phenylphenanthridinium bromide,3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide,phenanthridium 1553 PubChem CID: 68207 IUPAC Name: 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide SMILES: [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Dimidium bromide, ∽95%, MP Biomedicals™
CAS: 518-67-2 Molecular Formula: C20H18BrN3 Molecular Weight (g/mol): 380.29 MDL Number: MFCD00011757 InChI Key: MQOKYEROIFEEBH-UHFFFAOYSA-N Synonym: dimidium bromide,trypadine,3,8-diamino-5-methyl-6-phenylphenanthridinium bromide,phenanthridinium 1553,phenanthridinium compound 1553,phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide,2,7-diamino-10-methyl-9-phenylphenanthridinium bromide,3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide,phenanthridium 1553 PubChem CID: 68207 IUPAC Name: 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide SMILES: [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Dimidium Bromide, MP Biomedicals
CAS: 518-67-2 Molecular Formula: C20H18BrN3 Molecular Weight (g/mol): 380.29 MDL Number: MFCD00011757 InChI Key: MQOKYEROIFEEBH-UHFFFAOYSA-N Synonym: dimidium bromide,trypadine,3,8-diamino-5-methyl-6-phenylphenanthridinium bromide,phenanthridinium 1553,phenanthridinium compound 1553,phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide,2,7-diamino-10-methyl-9-phenylphenanthridinium bromide,3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide,phenanthridium 1553 PubChem CID: 68207 IUPAC Name: 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide SMILES: [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12