Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimidium Bromide, 97%, Spectrum™ Chemical
SDP

Catalog No. 18610255 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-255 500 mg Amber Glass Bottle
18-610-253 100 mg Amber Glass Bottle
18-610-250 1 g Amber Glass Bottle
18-610-252 100 g Amber Glass Bottle
18-610-251 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-610-255 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2499500MG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 518-67-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mg
Molecular Weight (g/mol) 380.31
Percent Purity 0.97
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Dimidium Bromide

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.