accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaOSOOSOOOSOOO

CAS RN 5182-30-9

IUPAC Name: trisodium naphthalene-1,3,6-trisulfonate
Synonyms: trisodium naphthalene trisulfonate
Molecular Weight (g/mol): 434.29
Molecular Formula: C10H5Na3O9S3
InChi Key: NJPKYOIXTSGVAN-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)