Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene-1,3,6-trisulfonic Acid, Na (Technical),MP Biomedicals

Catalog No. ICN214480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214480 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214480 Supplier MP Biomedicals, Inc Supplier No. 0521448050
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5182-30-9
Molecular Formula C10H5Na3O9S3
Molecular Weight (g/mol) 434.29
MDL Number MFCD00003989
InChI Key NJPKYOIXTSGVAN-UHFFFAOYSA-K
Synonym trisodium 1,3,6-naphthalenetrisulfonate, 1,3,6-naphthalenetrisulfonic acid, trisodium salt, naphthalene-1,3,6-trisulfonic acid trisodium salt, trisodium naphthalene-1,3,6-trisulfonate, sodium naphthalene-1,3,6-trisulfonate, trisodium 1,3,6-naphthalene trisulfonate, trisodium naphthalene-1,3,6-trisulphonate, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:3, 1,3,6-naphthalene trisulfonic acid trisodium salt, trisodium naphthalene trisulfonate
PubChem CID 78855
IUPAC Name trisodium naphthalene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Quantity 50 mg
Chemical Name or Material Naphthalene-1,3,6-Trisulfonic Acid, Na, Tech
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.