accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51851-79-7

IUPAC Name: bis(propan-2-yl) (prop-2-en-1-yl)boronate
Synonyms: diisopropyl prop-2-en-1-ylboronate
Molecular Weight (g/mol): 170.06
Molecular Formula: C9H19BO2
InChi Key: LWPLTONTMJTRJL-UHFFFAOYSA-N
SMILES: CC(C)OB(CC=C)OC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results