Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Allylboronate 95.0+%, TCI America™
SDP

Catalog No. D37385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37385G 5 g
D37381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37385G Supplier TCI America Supplier No. D37385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51851-79-7
Molecular Formula C9H19BO2
Molecular Weight (g/mol) 170.06
MDL Number MFCD15072157
InChI Key LWPLTONTMJTRJL-UHFFFAOYSA-N
Synonym Allylboronic Acid Diisopropyl Ester
PubChem CID 11435180
IUPAC Name bis(propan-2-yl) (prop-2-en-1-yl)boronate
SMILES CC(C)OB(CC=C)OC(C)C

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 170.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Allylboronate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.