accessibility menu, dialog, popup

UserName

Viewing profile as user:
O(R)(R)N(S)N(S)

CAS RN 519-02-8

IUPAC Name: (1R,2R,9S,17S)-7,13-diazatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadecan-6-one
Synonyms: matrine matrene, (+)- (5β)-isomer matrine sophoridine matrine, (5β)-isomer (+)-matrine (+)-matrene (5β)-matrine matridin-15-one (9CI) matrine, (5β)-
Molecular Weight (g/mol): 248.37
Molecular Formula: C15H24N2O
InChi Key: ZSBXGIUJOOQZMP-JLNYLFASSA-N
SMILES: O=C1CCC[C@@H]2[C@H]3CCCN4CCC[C@@H](CN12)[C@@H]34
Color 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)