CAS RN 519-09-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Benzoylecgonine Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
This certified reference standard is suitable for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.