accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 519-23-3

H3CHNCH3N

CAS RN 519-23-3

IUPAC Name: 5,11-dimethyl-6H-pyrido[4,3-b]carbazole
Synonyms: ellipticine elliptisine
Molecular Weight (g/mol): 246.31
Molecular Formula: C17H14N2
InChi Key: CTSPAMFJBXKSOY-UHFFFAOYSA-N
SMILES: CC1=C2C=CN=CC2=C(C)C2=C1NC1=C2C=CC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results