Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ellipticine

Catalog No. 32468810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-468-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-468-810MG Supplier MilliporeSigma™ Supplier No. 32468810MG
Add to Cart
Add to Cart

A cell-permeable antitumor alkaloid that acts as an inhibitor of topoisomerase II and acts as an intercalative agent that stimulates topoisomerase II-mediated DNA breakage.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Bright Yellow
Description 99% by HPLC
Target Proteins topoisomerase 2
CAS 519-23-3
Molecular Formula C17H14N2
Formula Weight 246.3g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.