accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 519017-62-0

IUPAC Name: N-(4-bromo-2-fluorophenyl)-4-chlorobenzamide
Synonyms: N-(4-bromo-2-fluorophenyl)-4-chlorobenzamide
Molecular Weight (g/mol): 328.57
Molecular Formula: C13H8BrClFNO
InChi Key: XLPWNTBBCDMYQV-UHFFFAOYSA-N
SMILES: FC1=C(NC(=O)C2=CC=C(Cl)C=C2)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results