accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSOOHHOOH

CAS RN 519054-53-6

IUPAC Name: 2-(dihydroxyboranyl)thiophene-3-carboxylic acid
Synonyms: 2-(dihydroxyboranyl)thiophene-3-carboxylic acid
Molecular Weight (g/mol): 171.96
Molecular Formula: C5H5BO4S
InChi Key: CGKJHZDZZNMEAP-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=CS1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results