Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4-Dihydroxy-5-propylphenyl)ethan-1-one, Tech., Thermo Scientific™

Catalog No. MO00639DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO00639DE 5 g
MO00639DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO00639DE Supplier Thermo Scientific Chemicals Supplier No. MO00639DE

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 519054-53-6
Molecular Formula C5H5BO4S
Molecular Weight (g/mol) 171.96
MDL Number MFCD03407366
InChI Key CGKJHZDZZNMEAP-UHFFFAOYSA-N
Synonym 2-dihydroxyboryl-3-thiophenecarboxylic acid, 3-carboxythiophene-2-boronic acid, 2-dihydroxyboranyl thiophene-3-carboxylic acid, 3-thiophenecarboxylicacid, 2-borono, 2dihydroxyboryl3thiophenecarboxylic acid, 2-borono-3-thiophenecarboxylic acid, 3-thiophenecarboxylic acid, 2-borono
PubChem CID 2794616
IUPAC Name 2-(dihydroxyboranyl)thiophene-3-carboxylic acid
SMILES OB(O)C1=C(C=CS1)C(O)=O

Specifications

Quantity 5 g
Formula Weight 171.97
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(Dihydroxyboryl)-3-thiophenecarboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.