accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CH3CCH3H3CNOCH3

CAS RN 519054-54-7

IUPAC Name: 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-benzoxazine
Molecular Weight (g/mol): 275.16
Molecular Formula: C15H22BNO3
InChi Key: QRAOZQGIUIDZQZ-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results