Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific™

Catalog No. CC13539DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13539DE 5 g Amber Glass Bottle
CC13539CB 250 mg Amber Glass Bottle
CC13539DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC13539DE Supplier Thermo Scientific Chemicals Supplier No. CC13539DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 519054-54-7
Molecular Formula C15H22BNO3
Molecular Weight (g/mol) 275.16
MDL Number MFCD04115377
InChI Key QRAOZQGIUIDZQZ-UHFFFAOYSA-N
Synonym 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-ylboronic acid pinacol ester, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2,-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-d ioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine
PubChem CID 2795301
IUPAC Name 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-benzoxazine
SMILES CN1CCOC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 275.15
Chemical Name or Material 4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.