accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HN

CAS RN 5192-23-4

IUPAC Name: 1H-indol-4-amine
Synonyms: 1H-indol-4-amine
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: LUNUNJFSHKSXGQ-UHFFFAOYSA-N
SMILES: NC1=C2C=CNC2=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)