Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoindole, 98%, Thermo Scientific™

Catalog No. AAL17792MC
Change view
Click to view available options
Quantity:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17792MC 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL17792MC Supplier Thermo Scientific Chemicals Supplier No. L17792MC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5192-23-4
Molecular Formula C8H8N2
Quantity 100mg
MDL Number MFCD01076559
Beilstein 114919
Sensitivity Air sensitive
Synonym 4-aminoindole, 1h-indol-4-ylamine, 4-amino indole, 4-amino-1h-indole, 1h-indol-4-yl-amine, 4-indolamine, indole-4-ylamine, 1h-indol-4-amin, 4-aminoindol, indol-4-ylamine
InChI Key LUNUNJFSHKSXGQ-UHFFFAOYSA-N
SMILES C1=CC2=C(C=CN2)C(=C1)N
IUPAC Name 1H-indol-4-amine
Molecular Weight (g/mol) 132.166
PubChem CID 583431
Formula Weight 132.17
Percent Purity 98%
Chemical Name or Material 4-Aminoindole
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 621-202-0
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.