accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5193-03-3

IUPAC Name: 2-chloro-6-hydrazinylpyridine
Synonyms: 2-chloro-6-hydrazinylpyridine
Molecular Weight (g/mol): 143.57
Molecular Formula: C5H6ClN3
InChi Key: HFTSPKIAXHZROA-UHFFFAOYSA-N
SMILES: NNC1=CC=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)