Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-hydrazinopyridine 98.0+%, TCI America™
SDP

Catalog No. C24635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24635G Supplier TCI America Supplier No. C24635G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent with 98.0% purity for research applications.

Chemical Identifiers

CAS 5193-03-3
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.57
MDL Number MFCD00214724
InChI Key HFTSPKIAXHZROA-UHFFFAOYSA-N
Synonym 2-chloro-6-hydrazinopyridine, 2-chloro-6-hydrazinylpyridine, 6-chloro-2-pyridyl hydrazine, 6-chloro-2-pyridylhydrazine, 2 1h-pyridinone, 6-chloro-, hydrazone, pubchem12983, acmc-1awi0, 2-hydrazino-6-chloropyridine, 6-chloro-2-hydrazinopyridine, 6-chloro-2-hydrazonopyridine
PubChem CID 320004
IUPAC Name (6-chloropyridin-2-yl)hydrazine
SMILES C1=CC(=NC(=C1)Cl)NN

Specifications

Melting Point 120°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-hydrazinopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.