accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51940-44-4

IUPAC Name: 8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid
Synonyms: pipemidic acid 吡哌酸
Molecular Weight (g/mol): 303.32
Molecular Formula: C14H17N5O3
InChi Key: JOHZPMXAZQZXHR-UHFFFAOYSA-N
SMILES: CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results