Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pipemidic Acid, MP Biomedicals™

Catalog No. ICN15626610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15626610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15626610 Supplier MP Biomedicals, Inc Supplier No. 0215626610

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51940-44-4
Molecular Formula C14H17N5O3
Molecular Weight (g/mol) 303.32
MDL Number MFCD00057291
InChI Key JOHZPMXAZQZXHR-UHFFFAOYSA-N
Synonym pipemidic acid, deblaston, pipram, palin, acido pipemidico, acide pipemidique, acidum pipemidicum, pipedac, pipemid, uromidin
PubChem CID 4831
ChEBI CHEBI:75250
IUPAC Name 8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid
SMILES CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1

Specifications

Quantity 10 g
Formula Weight 303.3
Physical Form Solid
Chemical Name or Material Pipemidic Acid
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.