accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCH3NCH3H3COCH3OH3COH3C

CAS RN 52-53-9

IUPAC Name: 2-(3,4-dimethoxyphenyl)-5-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}-2-(propan-2-yl)pentanenitrile
Synonyms: veraβ isoptin verpamil verapamil riker praecicor arpamyl rapam vera-sanorania civicor retard securon
Molecular Weight (g/mol): 454.61
Molecular Formula: C27H38N2O4
InChi Key: SGTNSNPWRIOYBX-UHFFFAOYNA-N
SMILES: COC1=CC=C(CCN(C)CCCC(C#N)(C(C)C)C2=CC=C(OC)C(OC)=C2)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results