Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verapamil hydrochloride, 99.9%, MP Biomedicals™

Catalog No. ICN19554505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19554505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19554505 Supplier MP Biomedicals, Inc Supplier No. 0219554505

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52-53-9
Molecular Formula C27H38N2O4
Molecular Weight (g/mol) 454.611
InChI Key SGTNSNPWRIOYBX-UHFFFAOYSA-N
Synonym verapamil, iproveratril, vasolan, dilacoran, isoptin, cordilox, calan, verapamilo, verapamilum, falicard
PubChem CID 2520
ChEBI CHEBI:77733
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC

Specifications

Color White
Assay Percent Range 0.999
Quantity 5 g
Formula Weight 491.1
Physical Form Powder
Chemical Name or Material Verapamil hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.