accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52-62-0

IUPAC Name: 1-methyl-1-[5-(1-methylpyrrolidin-1-ium-1-yl)pentyl]pyrrolidin-1-ium bis((2R,3R)-3-carboxy-2,3-dihydroxypropanoate)
Synonyms: pentolinium bitartrate pentapyrrolidium bitartrate tartrato de pentolonio pentolonium tartrate pendine pentolinium ditartrate recuryl pentolineum tartrate tensilest pentolonii tartras
Molecular Weight (g/mol): 538.59
Molecular Formula: C23H42N2O12
InChi Key: HSMKTIKKPMTUQH-WBPXWQEISA-L
SMILES: O[C@H]([C@@H](O)C([O-])=O)C(O)=O.O[C@H]([C@@H](O)C([O-])=O)C(O)=O.C[N+]1(CCCCC[N+]2(C)CCCC2)CCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results