Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentolinium Tartrate 99%, Thermo Scientific™

Catalog No. AC229941000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC229941000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC229941000 Supplier Thermo Scientific Chemicals Supplier No. 229941000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-62-0
Molecular Formula C23H42N2O12
Molecular Weight (g/mol) 538.591
InChI Key HSMKTIKKPMTUQH-RYOVOSQWSA-L
Synonym pentolinium ditartrate, lopac-p-3520, dsstox_cid_25542, dsstox_rid_80943, dsstox_gsid_45542, 1-methyl-1-5-1-methylpyrrolidin-1-ium-1-yl pentyl pyrrolidin-1-ium; 2s,3r-2,3,4-trihydroxy-4-oxobutanoate
PubChem CID 6604155
IUPAC Name 1-methyl-1-[5-(1-methylpyrrolidin-1-ium-1-yl)pentyl]pyrrolidin-1-ium;(2S,3R)-2,3,4-trihydroxy-4-oxobutanoate
SMILES C[N+]1(CCCC1)CCCCC[N+]2(CCCC2)C.C(C(C(=O)[O-])O)(C(=O)O)O.C(C(C(=O)[O-])O)(C(=O)O)O

Specifications

Color White
Infrared Spectrum Authentic
Quantity 100 mg
Merck Index 15, 7245
Formula Weight 538.58
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Pentolinium tartrate, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.