accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5205-34-5

IUPAC Name: decan-5-ol
Synonyms: decan-5-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: SZMNDOUFZGODBR-UHFFFAOYNA-N
SMILES: CCCCCC(O)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results