Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Decanol 96.0+%, TCI America™
SDP

Catalog No. D13815ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13815ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D13815ML Supplier TCI America Supplier No. D13815ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5205-34-5
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00039626
InChI Key SZMNDOUFZGODBR-UHFFFAOYSA-N
Synonym Amylbutylcarbinol, Butylpentylcarbinol, 5-Decyl Alcohol
PubChem CID 99868
IUPAC Name decan-5-ol
SMILES CCCCCC(CCCC)O

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 80°C
Quantity 5 mL
Formula Weight 158.29
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Decanol
EINECSNumber (2)-0217
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.