accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NHOCH3CH2

CAS RN 5205-93-6

IUPAC Name: N-[3-(dimethylamino)propyl]-2-methylprop-2-enamide
Synonyms: N-[3-(dimethylamino)propyl]-2-methylprop-2-enamide
Molecular Weight (g/mol): 170.26
Molecular Formula: C9H18N2O
InChi Key: GDFCSMCGLZFNFY-UHFFFAOYSA-N
SMILES: CN(C)CCCNC(=O)C(C)=C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)