Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylaminopropylmethylacrylamide, MP Biomedicals

Catalog No. ICN214502 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214502 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214502 Supplier MP Biomedicals, Inc Supplier No. 0521450280

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5205-93-6
Molecular Formula C9H18N2O
Molecular Weight (g/mol) 170.256
InChI Key GDFCSMCGLZFNFY-UHFFFAOYSA-N
Synonym n-3-dimethylamino propyl methacrylamide, dmapma, unii-63obq7bzds, 63obq7bzds, n-dimethylamino propyl methacrylamide, n-3-dimethylaminopropyl methacrylamide, n-3-dimethylamino propyl-2-methylprop-2-enamide, 2-propenamide, n-3-dimethylamino propyl-2-methyl
PubChem CID 78882
IUPAC Name N-[3-(dimethylamino)propyl]-2-methylprop-2-enamide
SMILES CC(=C)C(=O)NCCCN(C)C

Specifications

Quantity 100 g
Chemical Name or Material 3-Dimethylaminopropylmethylacrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.