accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52089-55-1

IUPAC Name: ethyl 2-hydroxyhexanoate
Synonyms: ethyl 2-hydroxyhexanoate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: MRYSSTRVUMCKKB-UHFFFAOYNA-N
SMILES: CCCCC(O)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results