Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl DL-2-hydroxycaproate, 97%, Thermo Scientific™

Catalog No. AC214030250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC214030250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC214030250 Supplier Thermo Scientific Chemicals Supplier No. 214030250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52089-55-1
Density 0.9600g/mL
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H16O3
Linear Formula CH3(CH2)3CH(OH)CO2C2H5
Quantity 25g
MDL Number MFCD00004583
Beilstein 03, 333
Synonym ethyl 2-hydroxycaproate, ethyl dl-2-hydroxycaproate, acmc-20miyg, hexanoic acid, 2-hydroxy-, ethyl ester, r, acmc-20al12, ethyl 2-dl-hydroxyhexanoate, ethyl 2-hydroxyhexanoate, ethyl +/--2-hydroxycaproate, 2-hydroxy-hexanoic acid ethyl ester
InChI Key MRYSSTRVUMCKKB-UHFFFAOYSA-N
SMILES CCCCC(C(=O)OCC)O
IUPAC Name ethyl 2-hydroxyhexanoate
Molecular Weight (g/mol) 160.213
PubChem CID 103589
Formula Weight 160.21
Chemical Name or Material Ethyl DL-2-hydroxycaproate, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.