accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOOFFFSOOOFFFSOOOLa

CAS RN 52093-26-2

IUPAC Name: lanthanum(3+) tritrifluoromethanesulfonate
Synonyms: lanthanum(3+) tritriflate
Molecular Weight (g/mol): 586.1
Molecular Formula: C3F9LaO9S3
InChi Key: WGJJZRVGLPOKQT-UHFFFAOYSA-K
SMILES: [La+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (4)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)