accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52093-28-4

IUPAC Name: samarium(3+) tritrifluoromethanesulfonate
Synonyms: samarium(3+) tritriflate
Molecular Weight (g/mol): 597.55
Molecular Formula: C3F9O9S3Sm
InChi Key: DDCWGUIPLGMBPO-UHFFFAOYSA-K
SMILES: [Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results