CAS RN 521-35-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
CERILLIANT™ Cannabinol Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
This analytical reference standard is suitable in LC/MS or GC/MS testing methods for applications in urine drug testing, clinical toxicology, pharmaceutical research, and forensic analysis.