accessibility menu, dialog, popup

UserName

Viewing profile as user:
O(S)OONH2HCl

CAS RN 52142-01-5

IUPAC Name: benzyl (2S)-2-amino-3-[4-(benzyloxy)phenyl]propanoate hydrochloride
Synonyms: benzyl (2S)-2-amino-3-[4-(benzyloxy)phenyl]propanoate hydrochloride
Molecular Weight (g/mol): 397.9
Molecular Formula: C23H24ClNO3
InChi Key: DYLKTBYVTWFYGQ-DKVUPULANA-N
SMILES: Cl.N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)