Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzyl-L-tyrosine benzyl ester hydrochloride, 95%, Thermo Scientific™

Catalog No. AAH6315606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6315606 5 g
AAH6315603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6315606 Supplier Thermo Scientific Chemicals Supplier No. H6315606

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52142-01-5
Molecular Formula C23H24ClNO3
Molecular Weight (g/mol) 397.90
MDL Number MFCD00153465
InChI Key DYLKTBYVTWFYGQ-DKVUPULANA-N
Synonym h-tyr bzl-obzl.hcl, h-tyr bzl-obzl hcl, o-benzyl-l-tyrosine benzyl ester hydrochloride, l-tyrosine, o-phenylmethyl-, phenylmethyl ester, hydrochloride, httyr bzl-obzl hcl, h-tyr bzl-obzlcl, h-tyr bzl-obzl??hcl, h-tyr bzl-obzl hydrochloride, benzyl o-benzyl-l-tyrosinate-hydrogen chloride 1/1, benzyl 2s-2-amino-3-4-benzyloxyphenyl propanoate hydrochloride
PubChem CID 45073233
IUPAC Name benzyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride
SMILES Cl.N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 397.89
Percent Purity 95%
Physical Form Solid
Chemical Name or Material O-Benzyl-L-tyrosine benzyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.