CAS RN 52147-67-8
IUPAC Name:
methyl 2-methanesulfinylacetate
Synonyms:
methyl 2-methanesulfinylacetate
Molecular Weight (g/mol):
136.17
Molecular Formula:
C4H8O3S
InChi Key:
DWNQOKWDJKLVEW-UHFFFAOYNA-N
SMILES:
COC(=O)CS(C)=O
1
–
1
of
1
results
1
Methyl (methylsulfinyl)acetate (Tech.), 80%, Thermo Scientific™
CAS: 52147-67-8 Molecular Formula: C4H8O3S Molecular Weight (g/mol): 136.165 InChI Key: DWNQOKWDJKLVEW-UHFFFAOYSA-N Synonym: methyl 2-methanesulfinylacetate,methyl methylsulfinyl acetate,methyl methylsulphinyl acetate,methyl methylsulfinylacetate,methylsulfinyl acetic acid methyl ester,methyl methylsulfinyl acetate tech.,acetic acid, methylsulfinyl-, methyl ester,acetic acid,2-methylsulfinyl-, methyl ester PubChem CID: 104097 IUPAC Name: methyl 2-methylsulfinylacetate SMILES: COC(=O)CS(=O)C