Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (methylsulfinyl)acetate (Tech.), 80%, Thermo Scientific™

Catalog No. AC336230010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC336230010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC336230010 Supplier Thermo Scientific Chemicals Supplier No. 336230010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52147-67-8
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.165
InChI Key DWNQOKWDJKLVEW-UHFFFAOYSA-N
Synonym methyl 2-methanesulfinylacetate, methyl methylsulfinyl acetate, methyl methylsulphinyl acetate, methyl methylsulfinylacetate, methylsulfinyl acetic acid methyl ester, methyl methylsulfinyl acetate tech., acetic acid, methylsulfinyl-, methyl ester, acetic acid,2-methylsulfinyl-, methyl ester
PubChem CID 104097
IUPAC Name methyl 2-methylsulfinylacetate
SMILES COC(=O)CS(=O)C

Specifications

Color Colorless
Density 1.25
Infrared Spectrum Authentic
Refractive Index 1.4840 to 1.4900 (20°C, 589nm)
Linear Formula CH3S(O)CH2CO2CH3
Quantity 1 g
Formula Weight 136.17
Percent Purity ≥75%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl (methylsulfinyl)acetate, tech., 80%
Show More Show Less
Hazard Category STENCH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.