accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3C(R)NOOHH3C

CAS RN 522-66-7

IUPAC Name: (R)-{5-ethyl-1-azabicyclo[2.2.2]octan-2-yl}(6-methoxyquinolin-4-yl)methanol
Synonyms: dihydroquinine
Molecular Weight (g/mol): 326.44
Molecular Formula: C20H26N2O2
InChi Key: LJOQGZACKSYWCH-ADMHDHIONA-N
SMILES: CCC1CN2CCC1CC2[C@H](O)C1=CC=NC2=CC=C(OC)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results