CAS RN 52205-73-9
IUPAC Name:
disodium (1S,3aS,3bR,9bS,11aS)-7-{[bis(2-chloroethyl)carbamoyl]oxy}-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl phosphate
Synonyms:
emcyt estracyt estramustine phosphate sodium estramustine sodium phosphate estramustine phosphate disodium
Molecular Weight (g/mol):
564.35
Molecular Formula:
C23H30Cl2NNa2O6P
InChi Key:
IIUMCNJTGSMNRO-VVSKJQCTSA-L
SMILES:
[Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC(=O)N(CCCl)CCCl)=CC=C34)[C@@H]1CC[C@@H]2OP([O-])([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Estramustine phosphate sodium
CAS: 52205-73-9 Molecular Formula: C23H30Cl2NNa2O6P